siRNA Details
| Gene Symbol: Afmid | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027827.3 | NP_082103.1 | 10 | 861 | ||
| GenBank mRNA | AF399717.1 | - | 10 | 856 | ||
| AK018563.1 | - | 8 | 797 | |||
| AY099479.1 | - | 10 | 856 | |||
| BC024452.1 | - | 10 | 832 | |||
| BC043309.1 | - | 10 | 827 | |||
| BC052507.1 | - | 6 | 486 | |||
| BC066780.1 | - | 10 | 840 | 30301832 | ||

Afmid
Genomic Map