siRNA Details
| Gene Symbol: Clmp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133733.4 | NP_598494.2 | 7 | 1393 | ||
| GenBank mRNA | AB040490.1 | - | 7 | 1310 | ||
| AK018527.1 | - | 2 | 384 | |||
| AK028983.1 | - | 7 | 1363 | |||
| AK033723.1 | - | 7 | 1395 | |||
| AK080481.1 | - | 7 | 1350 | |||
| AY259213.1 | - | 8 | 1022 | |||
| AY326421.1 | - | 7 | 1028 | |||
| BC026447.1 | - | 7 | 1219 | 4459717 | ||

Clmp
Genomic Map