siRNA Details
| Gene Symbol: Mcm9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027830.2 | NP_082106.2 | 10 | 1943 | ||
| GenBank mRNA | AK133212.1 | - | 3 | 357 | ||
| AK133345.1 | - | 3 | 1170 | |||
| BC062185.1 | - | 1 | 20 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mcm9| Gene Symbol: Mcm9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027830.2 | NP_082106.2 | 10 | 1943 | ||
| GenBank mRNA | AK133212.1 | - | 3 | 357 | ||
| AK133345.1 | - | 3 | 1170 | |||
| BC062185.1 | - | 1 | 20 | |||