siRNA Details
| Gene Symbol: Ceacam20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027839.2 | NP_082115.2 | 3 | 382 | ||
| GenBank mRNA | AK018613.1 | - | 3 | 379 | ||
| AK033581.2 | - | 3 | 383 | |||
| BC049361.1 | - | 3 | 363 | 6476373 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ceacam20| Gene Symbol: Ceacam20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027839.2 | NP_082115.2 | 3 | 382 | ||
| GenBank mRNA | AK018613.1 | - | 3 | 379 | ||
| AK033581.2 | - | 3 | 383 | |||
| BC049361.1 | - | 3 | 363 | 6476373 | ||