siRNA Details
| Gene Symbol: Myo1e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181072.3 | NP_851417.2 | 12 | 1574 | ||
| GenBank mRNA | AK029145.1 | - | 12 | 1576 | ||
| BB220783.2 | - | 2 | 217 | |||
| BC040786.1 | - | 13 | 1630 | |||
| BC051391.1 | - | 12 | 1556 | 5053090 | ||

Myo1e
Genomic Map