siRNA Details
| Gene Symbol: Myo1e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181072.3 | NP_851417.2 | 13 | 1643 | ||
| GenBank mRNA | AK029145.1 | - | 13 | 1645 | ||
| BB220783.2 | - | 3 | 286 | |||
| BC040786.1 | - | 14 | 1699 | |||
| BC051391.1 | - | 13 | 1625 | 5053090 | ||

Myo1e
Genomic Map