siRNA Details
| Gene Symbol: Krtap5-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027844.4 | NP_082120.3 | 1 | 1332 | ||
| GenBank mRNA | AK014785.1 | - | 2 | 799 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Krtap5-2| Gene Symbol: Krtap5-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027844.4 | NP_082120.3 | 1 | 1332 | ||
| GenBank mRNA | AK014785.1 | - | 2 | 799 | ||