siRNA Details
| Gene Symbol: Zgrf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_197997.2 | NP_932114.2 | 6 | 1530 | ||
| GenBank mRNA | BC079891.1 | - | 6 | 1530 | ||
| BC082599.1 | - | 6 | 1414 | |||
| BC085187.1 | - | 6 | 1386 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zgrf1| Gene Symbol: Zgrf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_197997.2 | NP_932114.2 | 6 | 1530 | ||
| GenBank mRNA | BC079891.1 | - | 6 | 1530 | ||
| BC082599.1 | - | 6 | 1414 | |||
| BC085187.1 | - | 6 | 1386 | |||