siRNA Details
| Gene Symbol: Zgrf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_197997.2 | NP_932114.2 | 15 | 3993 | ||
| GenBank mRNA | AK046429.1 | - | 10 | 1718 | ||
| AK052386.1 | - | 1 | 1682 | |||
| BC079891.1 | - | 14 | 3869 | |||
| BC082599.1 | - | 13 | 5031 | |||

Zgrf1
Genomic Map