siRNA Details
| Gene Symbol: Zgrf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_197997.2 | NP_932114.2 | 17 | 4347 | ||
| GenBank mRNA | AK046429.1 | - | 12 | 2072 | ||
| AK156938.1 | - | 2 | 340 | |||
| BC079891.1 | - | 16 | 4223 | |||
| BC082599.1 | - | 15 | 5385 | |||

Zgrf1
Genomic Map