siRNA Details
| Gene Symbol: Tmem248 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081394.1 | NP_001074863.1 | 2 | 315 | ||
| NM_027854.1 | NP_082130.1 | 2 | 377 | |||
| GenBank mRNA | AK002297.1 | - | 2 | 315 | ||
| AK142176.1 | - | 2 | 365 | |||
| AK171152.1 | - | 2 | 343 | |||
| AK171607.1 | - | 2 | 313 | |||
| BC033455.1 | - | 2 | 287 | 4018584 | ||
| CN526906.1 | - | 2 | 402 | |||

Tmem248
Genomic Map