siRNA Details
Gene Symbol: Tmem248 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081394.1 | NP_001074863.1 | 5, 6 | 984 | ||
NM_027854.1 | NP_082130.1 | 5, 6 | 1046 | |||
GenBank mRNA | AK002297.1 | - | 5, 6 | 984 | ||
AK142176.1 | - | 5, 6 | 1034 | |||
AK171152.1 | - | 5, 6 | 1012 | |||
BC033455.1 | - | 5, 6 | 956 | 4018584 |
