siRNA Details
| Gene Symbol: Acy3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF356878.1 | - | 8 | 1047 | ||
| AF356879.1 | - | 7 | 906 | |||
| AF375479.1 | - | 6 | 858 | |||
| AK002247.1 | - | 8 | 1046 | |||
| AK143782.1 | - | 8 | 1049 | |||
| AY169234.1 | - | 8 | 1046 | |||
| BC010795.1 | - | 6 | 1267 | 4221711 | ||

Acy3
Genomic Map