siRNA Details
| Gene Symbol: 0610010F05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027860.2 | NP_082136.2 | 19 | 2196 | ||
| GenBank mRNA | AK002465.1 | - | 5 | 313 | ||
| AK029538.1 | - | 19 | 2118 | |||
| AK129454.1 | - | 19 | 2163 | |||
| AK142320.1 | - | 4 | 222 | |||
| BC065122.1 | - | 20 | 2220 | |||
| BC072622.1 | - | 19 | 2187 | 6821718 | ||
| BC079876.1 | - | 19 | 2182 | 6848269 | ||

0610010F05Rik
Genomic Map