siRNA Details
| Gene Symbol: Brox | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027861.2 | NP_082137.1 | 13, 14 | 1366 | ||
| GenBank mRNA | AK036197.1 | - | 7, 8 | 701 | ||
| AK050525.1 | - | 13, 14 | 1337 | |||
| AK146863.1 | - | 12, 13 | 1226 | |||
| AK162247.1 | - | 13, 14 | 1336 | |||
| BC030340.1 | - | 13, 14 | 1372 | 3498026 | ||
| BC120692.1 | - | 12, 13 | 1214 | 40129615 | ||
| BC125527.1 | - | 12, 13 | 1214 | 40130042 | ||

Brox
Genomic Map