siRNA Details
| Gene Symbol: Gypc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001048207.1 | NP_001041672.1 | 3 | 843 | ||
| GenBank mRNA | AK002769.1 | - | 3 | 673 | ||
| AK082930.1 | - | 3 | 757 | |||
| AK152607.1 | - | 3 | 773 | |||
| AK159730.1 | - | 3 | 777 | |||
| AK162472.1 | - | 2 | 576 | |||
| AY234183.1 | - | 3 | 843 | |||
| BC027408.1 | - | 3 | 783 | 4952917 | ||

Gypc
Genomic Map