siRNA Details
| Gene Symbol: Cdc5l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152810.2 | NP_690023.1 | 10 | 1569 | ||
| GenBank mRNA | AK004547.2 | - | 10 | 1599 | ||
| AK146892.1 | - | 10 | 1389 | |||
| AK150562.1 | - | 10 | 1386 | |||
| AK150861.1 | - | 10 | 1386 | |||
| AK172950.1 | - | 10 | 1389 | |||
| BC003893.1 | - | 11 | 1504 | 3593535 | ||
| BC031480.1 | - | 10 | 1569 | 3498853 | ||

Cdc5l
Genomic Map