siRNA Details
| Gene Symbol: Arhgef3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289686.1 | NP_001276615.1 | 8 | 741 | ||
| NM_001289687.1 | NP_001276616.1 | 5 | 612 | |||
| NM_001289688.1 | NP_001276617.1 | 5 | 786 | |||
| NM_027871.2 | NP_082147.1 | 6 | 586 | |||
| GenBank mRNA | AK004600.2 | - | 6 | 557 | ||
| AK029872.1 | - | 9 | 771 | |||
| BC005517.1 | - | 5 | 583 | 3494309 | ||
| BC012262.1 | - | 5 | 564 | 4192378 | ||

Arhgef3
Genomic Map