siRNA Details
| Gene Symbol: Slc46a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027872.3 | NP_082148.1 | 3 | 1391 | ||
| GenBank mRNA | AK004609.1 | - | 3 | 1176 | ||
| AK156898.1 | - | 3 | 1393 | |||
| BC006902.1 | - | 3 | 1172 | 3599960 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc46a3| Gene Symbol: Slc46a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027872.3 | NP_082148.1 | 3 | 1391 | ||
| GenBank mRNA | AK004609.1 | - | 3 | 1176 | ||
| AK156898.1 | - | 3 | 1393 | |||
| BC006902.1 | - | 3 | 1172 | 3599960 | ||