siRNA Details
| Gene Symbol: Telo2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163661.1 | NP_001157133.1 | 8 | 1640 | ||
| NM_027880.2 | NP_082156.2 | 8 | 1551 | |||
| GenBank mRNA | AK004582.1 | - | 8 | 1421 | ||
| AK028658.1 | - | 8 | 1405 | |||
| AK045321.1 | - | 1 | 1087 | |||
| AK050773.1 | - | 8 | 1370 | |||
| AK084830.1 | - | 8 | 1421 | |||
| AK129189.1 | - | 8 | 1633 | |||
| AK143827.1 | - | 8 | 1641 | |||
| AK146963.1 | - | 8 | 1427 | |||
| AK154727.1 | - | 8 | 1453 | |||
| BC011077.1 | - | 8 | 1401 | 3984911 | ||
| BC080713.1 | - | 8 | 1580 | |||

Telo2
Genomic Map