siRNA Details
| Gene Symbol: Cic | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB093224.1 | - | 6 | 2168 | ||
| AF363690.1 | - | 11 | 2898 | |||
| AK004685.1 | - | 2 | 297 | |||
| AK082152.1 | - | 1 | 60 | |||
| AK147276.1 | - | 11 | 3531 | |||
| AK147535.1 | - | 12 | 5649 | |||
| BC035474.1 | - | 2 | 236 | |||
| BC040463.1 | - | 2 | 598 | |||
| BC058665.1 | - | 8 | 2416 | |||

Cic
Genomic Map