siRNA Details
| Gene Symbol: Aox3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023617.2 | NP_076106.2 | 10 | 966 | ||
| GenBank mRNA | AF172276.2 | - | 10 | 1042 | ||
| AK030109.1 | - | 6 | 506 | |||
| AK149434.1 | - | 10 | 927 | |||
| AY347569.1 | - | 10 | 843 | |||
| AY347570.1 | - | 10 | 843 | |||
| BB616838.1 | - | 6 | 505 | |||
| BC049853.1 | - | 10 | 918 | |||
| BC138876.1 | - | 10 | 945 | 8861898 | ||

Aox3
Genomic Map