siRNA Details
| Gene Symbol: Susd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001162913.1 | NP_001156385.1 | 8 | 1286 | ||
| NM_027890.5 | NP_082166.3 | 10 | 1646 | |||
| GenBank mRNA | AK004703.2 | - | 10 | 1616 | ||
| AK028817.1 | - | 10 | 1622 | |||
| AK033823.1 | - | 8 | 1259 | |||
| AK037059.1 | - | 10 | 1628 | |||
| AK037127.1 | - | 10 | 1597 | |||
| AK038944.1 | - | 8 | 1914 | |||
| AK042469.1 | - | 10 | 1630 | |||
| AK128959.1 | - | 10 | 1618 | |||
| AK167155.1 | - | 9 | 1828 | |||
| BC026837.1 | - | 10 | 1599 | 4222578 | ||

Susd2
Genomic Map