siRNA Details
| Gene Symbol: Lrwd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027891.4 | NP_082167.2 | 12 | 1616 | ||
| GenBank mRNA | AK004717.1 | - | 10 | 2578 | ||
| AK084948.1 | - | 12 | 1616 | |||
| AK085327.1 | - | 1 | 1647 | |||
| AK153976.1 | - | 12 | 1587 | |||
| AK154352.1 | - | 12 | 1558 | |||
| AK167020.1 | - | 12 | 1565 | |||
| AK172708.1 | - | 12 | 1584 | |||
| AK188535.1 | - | 2 | 75 | |||
| AK207344.1 | - | 4 | 323 | |||
| BC048397.1 | - | 12 | 1595 | 6396815 | ||

Lrwd1
Genomic Map