siRNA Details
| Gene Symbol: Ulk3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027895.1 | NP_082171.1 | 4 | 525 | ||
| GenBank mRNA | AK004783.1 | - | 4 | 509 | ||
| AK032030.1 | - | 2 | 146 | |||
| AK154642.1 | - | 4 | 438 | |||
| BC037093.1 | - | 4 | 546 | |||
| BC151153.1 | - | 4 | 451 | 9088047 | ||

Ulk3
Genomic Map