siRNA Details
| Gene Symbol: Coasy | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF521095.1 | - | 4 | 967 | ||
| AK004878.1 | - | 4 | 1027 | |||
| AK159405.1 | - | 4 | 1016 | |||
| AK159912.1 | - | 3 | 1316 | |||
| AK166894.1 | - | 3 | 1310 | |||
| AK170330.1 | - | 3 | 1325 | |||
| BC020046.1 | - | 3 | 437 | |||
| BC064753.1 | - | 3 | 955 | |||

Coasy
Genomic Map