siRNA Details
| Gene Symbol: Rgl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023622.4 | NP_076111.2 | 11 | 1365 | ||
| GenBank mRNA | AF237669.1 | - | 11 | 1324 | ||
| AF239661.1 | - | 11 | 1366 | |||
| AK004876.1 | - | 10 | 1098 | |||
| AK053550.1 | - | 11 | 1352 | |||
| AK131140.1 | - | 871 | ||||
| AK134654.1 | - | 1347 | ||||
| AK165023.1 | - | 10 | 1458 | |||
| AK165746.1 | - | 11 | 1359 | |||

Rgl3
Genomic Map