siRNA Details
| Gene Symbol: Rgl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023622.4 | NP_076111.2 | 14 | 1548 | ||
| GenBank mRNA | AF237669.1 | - | 14 | 1507 | ||
| AF239661.1 | - | 14 | 1549 | |||
| AK004876.1 | - | 11 | 1577 | |||
| AK053550.1 | - | 14 | 1535 | |||
| AK131140.1 | - | 1054 | ||||
| AK134654.1 | - | 1530 | ||||
| AK165023.1 | - | 11 | 1937 | |||
| AK165746.1 | - | 12 | 1839 | |||

Rgl3
Genomic Map