siRNA Details
| Gene Symbol: R3hdm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001168292.1 | NP_001161764.1 | 7 | 688 | ||
| NM_001168293.1 | NP_001161765.1 | 6 | 1003 | |||
| NM_027900.4 | NP_082176.4 | 6 | 1003 | |||
| GenBank mRNA | AK043545.1 | - | 6 | 1004 | ||
| AK122416.1 | - | 6 | 546 | |||
| BC043083.1 | - | 4 | 300 | |||
| BC053099.1 | - | 2 | 107 | |||
| BC064442.1 | - | 2 | 75 | |||
| BC117927.2 | - | 6 | 746 | 40097626 | ||

R3hdm2
Genomic Map