siRNA Details
| Gene Symbol: Map3k13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172821.3 | NP_766409.2 | 3 | 815 | ||
| GenBank mRNA | DQ480427.1 | - | 3 | 815 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Map3k13| Gene Symbol: Map3k13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172821.3 | NP_766409.2 | 3 | 815 | ||
| GenBank mRNA | DQ480427.1 | - | 3 | 815 | ||