siRNA Details
| Gene Symbol: Cyp2d40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023623.2 | NP_076112.2 | 1, 2 | 199 | ||
| GenBank mRNA | AK004984.1 | - | 1, 2 | 262 | ||
| BC061152.1 | - | 1, 2 | 199 | 30308952 | ||
| BC087906.1 | - | 1, 2 | 260 | 30306972 | ||

Cyp2d40
Genomic Map