siRNA Details
| Gene Symbol: Dhdh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027903.3 | NP_082179.1 | 5 | 673 | ||
| GenBank mRNA | AK005050.2 | - | 5 | 674 | ||
| AK154452.1 | - | 5 | 655 | |||
| BC031710.1 | - | 5 | 651 | |||
| BC116414.2 | - | 5 | 657 | 40103176 | ||

Dhdh
Genomic Map