siRNA Details
| Gene Symbol: Amdhd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027908.1 | NP_082184.1 | 5 | 740 | ||
| GenBank mRNA | AK005066.1 | - | 5 | 696 | ||
| AK090143.1 | - | 5 | 731 | |||
| BC022626.1 | - | 5 | 618 | |||
| BC043681.1 | - | 5 | 673 | |||
| BC055025.1 | - | 5 | 638 | |||
| BC118616.1 | - | 5 | 727 | 40107296 | ||
| BC121826.1 | - | 5 | 726 | 40107295 | ||

Amdhd1
Genomic Map