siRNA Details
| Gene Symbol: Klhdc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163729.2 | NP_001157201.2 | 6 | 924 | ||
| NM_027910.3 | NP_082186.2 | 7 | 1077 | |||
| GenBank mRNA | AB053465.1 | - | 7 | 948 | ||
| AJ278127.1 | - | 7 | 944 | |||
| AK004964.1 | - | 7 | 1054 | |||
| AK079082.1 | - | 7 | 955 | |||
| AK141673.1 | - | 6 | 909 | |||
| AK170623.1 | - | 7 | 949 | |||
| AK186202.1 | - | 1 | 106 | |||
| AK215007.1 | - | 1 | 36 | |||
| BC018154.1 | - | 7 | 1046 | 3983436 | ||

Klhdc3
Genomic Map