siRNA Details
| Gene Symbol: Shroom1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027917.3 | NP_082193.2 | 4 | 1294 | ||
| GenBank mRNA | AK004934.1 | - | 4 | 1221 | ||
| AK035462.1 | - | 3 | 2044 | |||
| AK134332.1 | - | 4 | 1222 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Shroom1| Gene Symbol: Shroom1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027917.3 | NP_082193.2 | 4 | 1294 | ||
| GenBank mRNA | AK004934.1 | - | 4 | 1221 | ||
| AK035462.1 | - | 3 | 2044 | |||
| AK134332.1 | - | 4 | 1222 | |||