siRNA Details
| Gene Symbol: Pdgfd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027924.2 | NP_082200.1 | 1 | 238 | ||
| GenBank mRNA | AF335583.1 | - | 1 | 55 | ||
| AK003359.2 | - | 1 | 240 | |||
| AK141551.1 | - | 1 | 267 | |||
| BC030896.1 | - | 1 | 189 | 4489485 | ||

Pdgfd
Genomic Map