siRNA Details
| Gene Symbol: Ints12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027927.3 | NP_082203.1 | 6 | 816 | ||
| GenBank mRNA | AK003874.1 | - | 6 | 828 | ||
| AK012829.1 | - | 6 | 827 | |||
| AK155162.1 | - | 6 | 828 | |||
| BC010657.1 | - | 4 | 488 | |||
| BC023801.1 | - | 6 | 816 | 5353058 | ||
| BC046996.1 | - | 6 | 810 | 6494551 | ||

Ints12
Genomic Map