siRNA Details
| Gene Symbol: Pitpnc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145823.2 | NP_665822.1 | 3 | 1046 | ||
| GenBank mRNA | AB077281.1 | - | 3 | 1027 | ||
| AB077282.1 | - | 3 | 324 | |||
| AK003844.1 | - | 2 | 294 | |||
| AK021206.1 | - | 3 | 259 | |||
| AK153625.1 | - | 3 | 1047 | |||
| AK171340.1 | - | 3 | 1073 | |||
| AK171918.1 | - | 3 | 1043 | |||
| AK172137.1 | - | 3 | 1047 | |||
| BC082333.1 | - | 3 | 878 | |||
| BC108351.1 | - | 3 | 906 | 6312193 | ||

Pitpnc1
Genomic Map