siRNA Details
| Gene Symbol: Pitpnc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145823.2 | NP_665822.1 | 2 | 955 | ||
| GenBank mRNA | AB077281.1 | - | 2 | 936 | ||
| AB077282.1 | - | 2 | 233 | |||
| AK003844.1 | - | 1 | 203 | |||
| AK021206.1 | - | 2 | 168 | |||
| AK153625.1 | - | 2 | 956 | |||
| AK171340.1 | - | 2 | 982 | |||
| AK171918.1 | - | 2 | 956 | |||
| AK172137.1 | - | 2 | 956 | |||
| BC082333.1 | - | 2 | 787 | |||
| BC108351.1 | - | 2 | 815 | 6312193 | ||

Pitpnc1
Genomic Map