siRNA Details
| Gene Symbol: Pitpnc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145823.2 | NP_665822.1 | 5, 6 | 1150 | ||
| GenBank mRNA | AB077281.1 | - | 5, 6 | 1131 | ||
| AB077282.1 | - | 5, 6 | 428 | |||
| AK003844.1 | - | 4, 5 | 398 | |||
| AK021206.1 | - | 5, 6 | 363 | |||
| AK153625.1 | - | 5, 6 | 1151 | |||
| AK171340.1 | - | 5, 6 | 1177 | |||
| AK171918.1 | - | 5, 6 | 1147 | |||
| BC082333.1 | - | 5, 6 | 982 | |||
| BC108351.1 | - | 5, 6 | 1010 | 6312193 | ||

Pitpnc1
Genomic Map