siRNA Details
| Gene Symbol: Chst13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027928.1 | NP_082204.1 | 3 | 979 | ||
| GenBank mRNA | AK004401.1 | - | 1 | 261 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Chst13| Gene Symbol: Chst13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027928.1 | NP_082204.1 | 3 | 979 | ||
| GenBank mRNA | AK004401.1 | - | 1 | 261 | ||