siRNA Details
| Gene Symbol: Plekhf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175175.4 | NP_780384.1 | 2 | 347 | ||
| GenBank mRNA | AK033666.1 | - | 2 | 341 | ||
| AK154634.1 | - | 2 | 338 | |||
| BC016134.1 | - | 2 | 288 | 4503757 | ||
| BC039276.1 | - | 2 | 298 | 4987735 | ||

Plekhf2
Genomic Map