siRNA Details
| Gene Symbol: Slc25a18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081048.2 | NP_001074517.1 | 2, 3 | 108 | ||
| GenBank mRNA | BC116828.1 | - | 2, 3 | 108 | 40126147 | |
| BC137603.1 | - | 2, 3 | 108 | 8860622 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc25a18| Gene Symbol: Slc25a18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081048.2 | NP_001074517.1 | 2, 3 | 108 | ||
| GenBank mRNA | BC116828.1 | - | 2, 3 | 108 | 40126147 | |
| BC137603.1 | - | 2, 3 | 108 | 8860622 | ||