siRNA Details
| Gene Symbol: Nup93 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172410.2 | NP_765998.1 | 15 | 1715 | ||
| GenBank mRNA | AK010450.1 | - | 14 | 1666 | ||
| AK028864.1 | - | 15 | 1716 | |||
| AK045380.1 | - | 1574 | ||||
| AK129055.1 | - | 13 | 2059 | |||
| AK167105.1 | - | 14 | 1666 | |||
| AK169194.1 | - | 12 | 1436 | |||
| AK169693.1 | - | 14 | 1685 | |||
| BC023140.1 | - | 14 | 1661 | 3991335 | ||

Nup93
Genomic Map