siRNA Details
| Gene Symbol: Nup93 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172410.2 | NP_765998.1 | 16, 17 | 1831 | ||
| GenBank mRNA | AK010450.1 | - | 15, 16 | 1782 | ||
| AK028864.1 | - | 16, 17 | 1832 | |||
| AK045380.1 | - | 1690 | ||||
| AK129055.1 | - | 14, 15 | 2175 | |||
| AK167105.1 | - | 15, 16 | 1782 | |||
| AK169194.1 | - | 13, 14 | 1552 | |||
| AK169693.1 | - | 15, 16 | 1801 | |||
| BC023140.1 | - | 15, 16 | 1777 | 3991335 | ||

Nup93
Genomic Map