siRNA Details
| Gene Symbol: Wdr34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008498.2 | NP_001008498.2 | 7 | 1203 | ||
| GenBank mRNA | AK014304.1 | - | 7 | 1202 | ||
| BC085113.1 | - | 7 | 1159 | 30730999 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Wdr34| Gene Symbol: Wdr34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008498.2 | NP_001008498.2 | 7 | 1203 | ||
| GenBank mRNA | AK014304.1 | - | 7 | 1202 | ||
| BC085113.1 | - | 7 | 1159 | 30730999 | ||