siRNA Details
Gene Symbol: Pdilt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027943.1 | NP_082219.1 | 9 | 1431 | ||
GenBank mRNA | AK005692.1 | - | 9 | 1431 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Pdilt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027943.1 | NP_082219.1 | 9 | 1431 | ||
GenBank mRNA | AK005692.1 | - | 9 | 1431 |