siRNA Details
| Gene Symbol: 1700007B14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164235.1 | NP_001157707.1 | 13, 14 | 1838 | ||
| NM_027944.1 | NP_082220.1 | 12, 13 | 1720 | |||
| GenBank mRNA | AK005693.1 | - | 12, 13 | 1721 | ||
| AK077102.1 | - | 14, 15 | 1577 | |||
| BC117767.1 | - | 13, 14 | 1741 | 40090978 | ||
| BC117768.1 | - | 12, 13 | 1623 | 40090984 | ||

1700007B14Rik
Genomic Map