siRNA Details
| Gene Symbol: Dcaf7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027946.3 | NP_082222.1 | 1 | 370 | ||
| GenBank mRNA | AK005902.1 | - | 1 | 364 | ||
| AK047435.1 | - | 1 | 372 | |||
| AK080823.1 | - | 1 | 343 | |||
| AK137813.1 | - | 1 | 334 | |||
| AK154757.1 | - | 1 | 361 | |||
| AK165674.1 | - | 1 | 361 | |||
| BB646282.1 | - | 1 | 371 | |||
| BC048165.1 | - | 1 | 286 | 6409531 | ||
| BC048722.1 | - | 1 | 289 | 6504571 | ||

Dcaf7
Genomic Map