siRNA Details
| Gene Symbol: Shcbp1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033162.2 | NP_001028334.1 | 5 | 922 | ||
| GenBank mRNA | AK005886.1 | - | 5 | 919 | ||
| AK161274.1 | - | 5 | 923 | |||
| AK171525.1 | - | 5 | 960 | |||
| BC113157.1 | - | 5 | 969 | 40058375 | ||

Shcbp1l
Genomic Map